Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium periodate
Sodium periodate
Sodium periodate
:
Merck
Sodium periodate
:
Merck
:
7790-28-5
:
:
NaIO₄
:
MFCD00003534
:
13,08714
:
300 °C (1013 hPa)
:
175 °C
:
3.87 g/cm³ (20 °C)
:
Ambient
:
232-197-6
:
5.1 (8),I
:
213.89 g/mol
:
3085
:
GHS03
:
GHS05
:
GHS08
:
GHS09
msds_106596
ProductPage_106596
ProductPage_106596