Sodium periodate 99.8+% ACS
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:7790-28-5
:
Formel:NaIO₄
MDL-nummer:MFCD00003534
Merck Index:13,08714
Kokande Pt:300 °C (1013 hPa)
Smält Pt:300 °C (dec.)
Densitet:3,87 g/cm³ (20 °C)
Förvaringstemperatur:Ambient
EINECS:232-197-6
ADR:5.1,I
:213.89 g/mol
FN:3085
:GHS03
:GHS05
:GHS08
:GHS09
Specifikationstestresultat
- Appearance:White Crystalline powder
- Titration Iodometric:99.8 to 100.3 % (on dried basis)
- Other halogens (as Cl):≤0.02 %
- Manganese (Mn):≤3 ppm