

Lithium fluoride ≥99.85% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-24-4
:
Formula:FLi
MDL Number:MFCD00011090
:25.94 g/mol
Merck Index:14,5531
Boiling Pt:1680 °C
Melting Pt:845 °C
Density:2.635
Storage Temperature:Ambient
EINECS:232-152-0
UN:3288
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together