Sodium chloride, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,8599
Boiling Pt:1413 °C
Melting Pt:801 °C
Density:2.165
:ampouled under argon
Storage Temperature:Ambient
EINECS:231-598-3