
Lithium chloride monohydrate ≥99.95% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16712-20-2
:85144-11-2
Formula:ClLi·1H₂O
MDL Number:MFCD00149764
:60.41 g/mol
Merck Index:14,5528
Boiling Pt:1382 °C (1013 hPa)
Melting Pt:>98 °C (-H₂O)
Density:1.78
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together