Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid ≥85%, CMOS for microelectronic, J.T.Baker®
Corrosive
Irritant
Orthophosphoric acid ≥85%, CMOS for microelectronic, J.T.Baker®
Catalog # 0274-11
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Orthophosphoric acid ≥85%, CMOS for microelectronic, J.T.Baker®
Catalog # 0274-11
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    700 lb
  • Seq:
    7365

Specifications

Specification Test Results

  • For Microelectronic Use:
    85.0 - 87.0 %
  • Store at temperatures above 22°C (72°F):
    ≤10
  • Assay (H₃PO₄) (by acidimetry):
    85.0 - 87.0 %
  • Color (APHA):
    ≤10
  • Specific Gravity at 60°/60°F:
    1.691 - 1.710
  • Reducing Substances:
    Passes Test
  • Volatile Acids (µeq/g):
    ≤0.16
  • Chloride (Cl):
    ≤1.0 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Sulfate (SO₄):
    ≤12 ppm
  • Trace Impurities - Aluminum (Al):
    ≤0.500 ppm
  • Trace Impurities - Antimony (Sb):
    ≤10.000 ppm
  • Trace Impurities - Arsenic (As):
    ≤0.050 ppm
  • Trace Impurities - Calcium (Ca):
    ≤1.500 ppm
  • Trace Impurities - Chromium (Cr):
    ≤0.200 ppm
  • Trace Impurities - Cobalt (Co):
    ≤0.050 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.050 ppm
  • Trace Impurities - Gold (Au):
    ≤0.300 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm
  • Trace Impurities - Lead (Pb):
    ≤0.300 ppm
  • Trace Impurities - Lithium (Li):
    ≤0.100 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤0.20 ppm
  • Trace Impurities - Manganese (Mn):
    ≤0.100 ppm
  • Trace Impurities - Nickel (Ni):
    ≤0.200 ppm
  • Trace Impurities - Potassium (K):
    ≤1.500 ppm
  • Trace Impurities - Sodium (Na):
    ≤2.500 ppm
  • Trace Impurities - Strontium (Sr):
    ≤0.100 ppm
  • Trace Impurities - Titanium (Ti):
    ≤0.300 ppm
  • Trace Impurities - Zinc (Zn):
    ≤2.000 ppm

Specification Test Results

About this item

 :
 :98 g/mol
Merck Index:14,08596
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
REACH:01-2119485498-19
 :GHS05
 :GHS07
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340