Order Entry
Sweden
ContactUsLinkComponent
dinatriumtetraborat, vattenfri ≥98%, GPR RECTAPUR®
Health
dinatriumtetraborat, vattenfri ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
dinatriumtetraborat, vattenfri ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Water:
    Max. 1.0%
  • Sulphate (SO4):
    Max. 150 ppm
  • Fe (Iron):
    Max. 40 ppm

Specification Test Results