
di-Sodium tetraborate
Catalog # MOLEM65439963
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:201.22 g/mol
Merck Index:12,08733
Boiling Pt:1575 °C (1013 hPa)
Melting Pt:742 °C
Density:2.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
CAS Number:1330-43-4
Formula:Na₂B₄O₇
MDL Number:MFCD00081185