Order Entry
Sweden
ContactUsLinkComponent
di-Sodium tetraborate, Suprapur® for trace analysis, Supelco®
Health
Irritant
di-Sodium tetraborate, Suprapur® for trace analysis, Supelco®
Catalog # 1.06309.0025
Supplier:  Merck
CAS Number:  
Health
Irritant
di-Sodium tetraborate, Suprapur® for trace analysis, Supelco®
Catalog # 1.06309.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    25.6 g/l (20 °C)
  • Melting Point:
    742 °C
  • Molar Mass:
    201.21 g/mol
  • Bulk Density:
    700 kg/m3
  • Boiling Point:
    1575 °C (decomposition)
  • Vapor Pressure:
    7.3 hPa (1200 °C)
  • Density:
    2.37 g/cm3 (20 °C)
  • pH:
    9.2 (25 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :201.22 g/mol
Merck Index:12,08733
Boiling Pt:1575 °C (1013 hPa)
Melting Pt:742 °C
Density:2.38 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
 :GHS08
 :GHS07
CAS Number:1330-43-4
Formula:Na₂B₄O₇
MDL Number:MFCD00081185