1,2-Benzisothiazol-3(2H)-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2634-33-5
:
Formula:C₇H₅NOS
MDL Number:MFCD00044001
:151.19 g/mol
EINECS:220-120-9
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:155.0 to 160.0 deg-C