Bis(2-ethylhexyl)adipate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:103-23-1
:
Formula:C₂₂H₄₂O₄
MDL Number:MFCD00009496
:370.57 g/mol
Flash Pt:205°C
EINECS:203-090-1
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %