Order Entry
Sweden
ContactUsLinkComponent
Sodium chloride 5 mol/l (5 N) in aqueous solution
Sodium chloride 5 mol/l (5 N) in aqueous solution
Catalog # FFCHFF63459010
Supplier:  FF CHEMICALS
CAS Number:  
Sodium chloride 5 mol/l (5 N) in aqueous solution
Catalog # FFCHFF63459010
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477