Order Entry
Sweden
ContactUsLinkComponent
 
Corrosive
Potassium hydroxide 2 mol/l (2 N) in aqueous solution
Catalog # FFCHFF0690050
Supplier:  FF CHEMICALS
CAS Number:  
Corrosive
Potassium hydroxide 2 mol/l (2 N) in aqueous solution
Catalog # FFCHFF0690050
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553