1,2-Benzisothiazol-3(2H)-one
Catalog # EHERC10536600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:151.19 g/mol
EINECS:220-120-9
CAS Number:2634-33-5
Formula:C₇H₅NOS
MDL Number:MFCD00044001