Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
2-Chloro-4-formylpyridine
2-Chloro-4-formylpyridine ≥97%
2-Chloro-4-formylpyridine ≥97%
:
Thermo Fisher Scientific
2-Chloro-4-formylpyridine ≥97%
:
Thermo Fisher Scientific
:
101066-61-9
:
:
C₆H₄ClNO
:
MFCD06651557
:
141.56 g/mol
:
46…49 °C
:
GHS07
:
Ambient
msds_ALFAH50098
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years