Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Scandium (III) trifluoromethanesulphonate
Scandium(III) trifluoromethanesulphonate 95%
Scandium(III) trifluoromethanesulphonate 95%
:
Thermo Fisher Scientific
Scandium(III) trifluoromethanesulphonate 95%
:
Thermo Fisher Scientific
:
144026-79-9
:
:
C₃F₉O₉S₃Sc
:
MFCD00192433
:
492.17 g/mol
:
300 °C
:
Ambient
:
3261
:
8,III
:
GHS05
Appearance:
White Powder
Infrared spectrum:
Conforms
Elemental Analysis:
≥94 %
ACRO36389