Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin (II) trifluoromethanesulphonate
Tin(II) trifluoromethanesulphonate 98%
Tin(II) trifluoromethanesulphonate 98%
:
Thermo Fisher Scientific
Tin(II) trifluoromethanesulphonate 98%
:
Thermo Fisher Scientific
:
62086-04-8
:
:
C₂F₆O₆S₂Sn
:
MFCD00191251
:
416.85 g/mol
:
>300 °C
:
Ambient
:
3261
:
8,III
:
GHS05
Appearance:
White to yellow Powder
Infrared spectrum:
Conforms
Assay:
≥97.5 % (ex Sn)
ACRO31253