Brij® L23 MBAL-PA-(MV)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-92-0
:
Formula:(C₂₀H₄₂O₅)n
MDL Number:MFCD00043063
Boiling Pt:>100 °C (760 mmHg)
Melting Pt:33…40 °C
Flash Pt:>149 °C
Storage Temperature:Ambient
EINECS:500-002-6
:GHS07
Specification Test Results
- Appearance:White Waxy pellets
- Infrared spectrum:Conforms
- Water:≤1 %
- Antioxidant:without antioxidant
- Impurity:≤10 ppm free Dioxane
- Acid value:≤5 mg KOH/g
- Hydroxyl value:40 to 60 mg KOH/g