Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
Triethyloxonium tetrafluoroborate 1 M in methylene chloride
 
undefined
 
 
 
Triethyloxonium tetrafluoroborate 1 M in methylene chloride

 

  • Appearance:
    Clear colorless to yellow to light brown Solution
  • Strength:
    0.9 to 1.1 M
  • Melting point:
    148 to 153 °C
  • Loss on drying:
    ≤0.5 % (1 g, 105 °C, 1 h)
  • HPLC:
    ≥97.5 %