Quinoline for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:91-22-5
:
Formula:C₉H₇N
MDL Number:MFCD00006736
:129.16 g/mol
Merck Index:13,08160
Boiling Pt:238 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.095 g/cm³ (20 °C)
EINECS:202-051-6
UN:2656
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:107 °C (closed cup)
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:6 g/l (20 °C)
- Melting Point:-15 °C
- Molar Mass:129.16 g/mol
- Boiling Point:237 - 238 °C (1013 hPa)
- Vapor Pressure:0.08 hPa (20 °C)
- Flash Point:107 °C
- Refractive Index:1.6268 (20 °C, 589 nm)
- Explosion Limit:1.2 - 7 %(V)
- Density:1.09 g/cm3 (20 °C)
- pH:7.3 (5 g/l, H2O, 20 °C)
- Ignition Point:480 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials