Order Entry
Sweden
ContactUsLinkComponent
3,5-Xylenol
3,5-Xylenol
Catalog # EHERC12735000
Supplier:  EHRENSTORFER
CAS Number:  
3,5-Xylenol
Catalog # EHERC12735000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :122.17 g/mol
Merck Index:12,10215
Boiling Pt:222 °C (1013 hPa)
Melting Pt:61…64 °C
Density:0.968 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-606-5
UN:2261
ADR:6.1,II
 :GHS06
 :GHS05
CAS Number:108-68-9
Formula:C₈H₁₀O
MDL Number:MFCD00002307