Order Entry
Sweden
ContactUsLinkComponent
2,6-Xylidine for synthesis, Sigma-Aldrich®
Health
Irritant
Environment
2,6-Xylidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
Environment
2,6-Xylidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    13 g/l (20 °C)
  • Melting Point:
    10 - 12 °C
  • Molar Mass:
    121.18 g/mol
  • Boiling Point:
    216 °C (1013 hPa)
  • Vapor Pressure:
    0.2 hPa (20 °C)
  • Flash Point:
    92 °C
  • Refractive Index:
    1.5601 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 6.9 %(V)
  • Density:
    0.98 g/cm3 (20 °C)
  • pH:
    12.5 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    490 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number