Order Entry
Sweden
ContactUsLinkComponent
 
 
Metanil yellow 70% (dye content), Sigma-Aldrich®
  202029-100G
 :  
undefined
 
Metanil yellow 70% (dye content), Sigma-Aldrich®
  202029-100G
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Appearance (colour):
    Gold to Orange
  • Appearance (form):
    Powder
  • pH Transition:
    pH 1.2 dark red or pink
    PH 2.7 yellow

 

 

 :
 :375.38 g/mol
 :M-Clarity™ quality level = MQ200
 :209-608-2
 :GHS05
 :Room Temperature
 :587-98-4
 :C₁₈H₁₄N₃NaO₃S
 :MFCD00007486