Order Entry
Sweden
ContactUsLinkComponent
 
Hydrochloric acid 0.25 mol/l (0.25 N) volumetric solution
Catalog # REGEH20255
Supplier:  REAGECON
CAS Number:  
Hydrochloric acid 0.25 mol/l (0.25 N) volumetric solution
Catalog # REGEH20255
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324