Order Entry
Sweden
ContactUsLinkComponent
 
Urea, Pierce™
Catalog # PIER29700
CAS Number:  
undefined
Urea, Pierce™
Catalog # PIER29700
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:13,09935
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
 :60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022