Order Entry
Sweden
ContactUsLinkComponent
Hydrochloric acid 40 - 60 g/l
Hydrochloric acid 40 - 60 g/l
Catalog # FFCHFF24015000
Supplier:  FF CHEMICALS
CAS Number:  
Hydrochloric acid 40 - 60 g/l
Catalog # FFCHFF24015000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Boiling Pt:∼100 °C (1013 hPa)
Density:1.02…1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,III
 :36.46 g/mol
EINECS:231-595-7
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324