


3-Aminobenzylamin 97%
:4403-70-7
:
:C₇H₁₀N₂
:MFCD00078355
:122.17 g/mol
:36…39 °C
:Omgivande
:412-230-2
:3259
:8,III
:GHS07
:GHS05
:GHS09
- Appearance:White to light brown Low melting solid or liquid
- Infrared spectrum:Conforms
- Melting point:38 to 44 °C
- GC:≥96.0 %