Order Entry
Sweden
ContactUsLinkComponent
4-Isopropylphenol 98%
4-Isopropylphenol 98%
Certificates
undefined
4-Isopropylphenol 98%

Specification Test Results

  • Appearance:
    White to pink to light brown Powder or crystals or fused solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 63 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information