

4-tert-Butylphenol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-54-4
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00002367
:150.22 g/mol
Merck Index:14,1585
Boiling Pt:236…238 °C
Melting Pt:97…101 °C
Density:0.908
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
EINECS:202-679-0
UN:3077
ADR:9,III
:GHS05
:GHS07