α-Chloro-4-nitrotoluene
Glass bottle for solids, 100 g
Catalog # MOLE46797071-100G
Marketsource
Supplier: Molekula
α-Chloro-4-nitrotoluene
Catalog # MOLE46797071-100G
Supplier: Molekula
Marketsource
CAS Number: 100-14-1
Jump to:
Product Details & Documents
Synonyms: 4-Nitrobenzyl chloride, 4-Nitrobenzylchloride, α-Chloro-p-nitrotoluene
- CAS Number:100-14-1
- Formula:C₇H₆ClNO₂
- MDL Number:MFCD00007374
- Molecular Weight:171.58 g/mol
- Boiling Pt:293 °C (1013 hPa)
- Melting Pt:68…70 °C
- Density:1.5647 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-822-7
- UN:3261
- ADR:8,II
- :Density: 1.5647 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



