Order Entry
Sweden
ContactUsLinkComponent
 
 
 
α-Chloro-3-fluorotoulene for synthesis
  8.18419.0010
 :  Merck
 :  
 
 
α-Chloro-3-fluorotoulene for synthesis
  8.18419.0010
 :  Merck
 :  

 

  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Solubility:
    40 g/l (20 °C)
  • Molar Mass:
    144.57 g/mol
  • Boiling Point:
    176 - 177 °C (1013 hPa)
  • Flash Point:
    58 °C
  • Density:
    1.21 g/cm3 (20 °C)

 

 

 :
 :144.58 g/mol
 :67 °C (1013 hPa)
 :1.21 g/cm³ (20 °C)
 :8 (3),II
 :M-Clarity™ quality level = MQ200
 :207-264-8
 :2920
 :GHS02
 :GHS05
 :Ambient
 :456-42-8
 :C₇H₆ClF
 :MFCD00000904