Order Entry
Sweden
ContactUsLinkComponent
Benzyl chloride 99% stabilised
Toxic
Health
Corrosive
Benzyl chloride 99% stabilised
Certificates
undefined
Toxic
Health
Corrosive
Benzyl chloride 99% stabilised

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5370 to 1.5390 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Stabilizer:
    0.25 % propylene oxide

Specification Test Results

Related Information