Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propiophenone
Propiophenone ≥99%
Propiophenone ≥99%
:
Thermo Fisher Scientific
Propiophenone ≥99%
:
Thermo Fisher Scientific
:
93-55-0
:
:
C₆H₅C(O)CH₂CH₃
:
MFCD00009309
:
134.18 g/mol
:
14,7830
:
217…218 °C
:
17…18 °C
:
1.008
:
87 °C (188 °F)
:
Ambient
:
202-257-6
msds_ALFAA15140
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years