Phenol
Catalog # PROOCHE110
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
:94.11 g/mol
Merck Index:12,07390
Boiling Pt:181.7 °C (1013 hPa)
Melting Pt:40.8 °C
Density:1.0722 g/cm³ (20 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
REACH:01-2119471329-32
:GHS06
:GHS05
:GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143