Order Entry
Sweden
ContactUsLinkComponent
Phenol 99.0-100.5%, detached crystals, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Phenol 99.0-100.5%, detached crystals, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Phenol 99.0-100.5%, detached crystals, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Assay (GC):
    Min. 99.5 %
  • Acidity:
    Passes test Ph.Eur.
  • Appearance of solution:
    Passes test Ph.Eur.
  • Clarity of solution:
    Passes test ACS
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Identification C:
    Passes test Ph.Eur.
  • Freezing point:
    Min. 40.5 °C
  • o-Cresol:
    Max. 0.05 %
  • m-Cresol:
    Max. 0.05 %
  • p-Cresol:
    Max. 0.05 %
  • Evaporation residue:
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Water:
    Max. 0.2 %
  • Cl (Chloride):
    Max. 5 ppm
  • Cu (Copper):
    Max. 0.1 ppm
  • Fe (Iron):
    Max. 1 ppm
  • K (Potassium):
    Max. 10 ppm
  • Na (Sodium):
    Max. 10 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.5 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results