Phenol 99.0-100.5%, detached crystals Ph. Eur.
:108-95-2
:
:C₆H₅OH
:MFCD00002143
:94.11 g/mol
:181.8 °C (1013 hPa)
:40.8 °C
:1.06 g/cm³ (20 °C)
:81 °C (closed cup)
:Ambient
:203-632-7
:1671
:6.1,II
:01-2119471329-32
:GHS06
:GHS05
:GHS08
- Assay:99.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Identification C:Passes test
- Appearance of solution:Passes test
- Acidity:Passes test
- Freezing point:Min. 39.5 °C
- Residue on evaporation:Max. 0.05 %
- Residual solvents:Passes test
Phenol 99 - 100,5%, Detached Crystals Ph. Eur.
Additional Product Details
SDS_EN_CH_VWRC20596.pdf SDS_EN_DE_VWRC20596.pdf SDS_EN_NL_VWRC20596.pdf SDS_EN_UK_VWRC20596.pdf sds_VWRC20596
SDS
spec_VWRC20596.297 spec_VWRC20596.297 spec_VWRC20596.468
Product specification
VWR Chemicals for Biopharma and Pharma Production