Order Entry
Sweden
ContactUsLinkComponent
1-Aminocyclopropanecarboxylic acid, white powder
1-Aminocyclopropanecarboxylic acid, white powder
Catalog # ICNA0215034883
Supplier:  MP Biomedicals
CAS Number:  
1-Aminocyclopropanecarboxylic acid, white powder
Catalog # ICNA0215034883
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard

Specification Test Results

About this item

 :
 :101.11 g/mol
Melting Pt:255…262 °C (dec., lit.)
 :GHS07
Storage Temperature:Freezer
CAS Number:22059-21-8
Formula:C₄H₇NO₂
MDL Number:MFCD00009944