Thiomorpholine
Glass bottle for solids, 5 g
Catalog # MOLEM67046947
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:123-90-0
- Formula:C₄H₉NS
- MDL Number:MFCD00005974
- Molecular Weight:103.19 g/mol
- EINECS:204-660-2
- UN:3267
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



