
(S)-(-)-1-(1-Naphthyl)ethylamine ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10420-89-0
:
Formula:C₁₂H₁₃N
MDL Number:MFCD00064179
:171.24 g/mol
Boiling Pt:125°C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 99.0 %
- Optical purity (GC):min. 98.0 ee%