Order Entry
Sweden
ContactUsLinkComponent
Indol-3-ylacetonitrile
Indol-3-ylacetonitrile
Catalog # EHERC14300000
Supplier:  EHRENSTORFER
CAS Number:  
Indol-3-ylacetonitrile
Catalog # EHERC14300000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :156.19 g/mol
EINECS:212-232-1
UN:3439
ADR:6.1,III
CAS Number:771-51-7
Formula:C₁₀H₈N₂
MDL Number:MFCD00005628