Indol-3-ylacetonitrile
Catalog # MOLE31916067-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:156.19 g/mol
EINECS:212-232-1
UN:3439
ADR:6.1,III
CAS Number:771-51-7
Formula:C₁₀H₈N₂
MDL Number:MFCD00005628