


O-Dianisidine dihydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20325-40-0
:
Formula:C₁₄H₁₈Cl₂N₂O₂
MDL Number:MFCD00012488
:317.21
Melting Pt:268 °C (dec.)
Storage Temperature:Ambient
EINECS:243-737-5
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White to beige to grey to purple-grey Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Titration with HClO4:≥97.5 % (on dry substance)