Mycophenolic acid
Catalog # MOLEM10127541
Supplier: Molekula
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:320.34 g/mol
EINECS:246-119-3
:GHS07
Storage Temperature:Refrigerator
CAS Number:24280-93-1
Formula:C₁₇H₂₀O₆
MDL Number:MFCD00036814