Organic reference standard, Picric acid 10 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERL17890500AL
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:88-89-1
- Description:Picric acid 10 µg/ml in acetonitrile
- Formula:C₆H₃N₃O₇
- Description:Picric acid