
6-Chloronicotinic acid, Supelco®
Catalog # 68678-100MG
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:100 mg
- Seq:2440
Specifications
About this item
:
:157.56 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:188…189 °C
EINECS:226-201-5
:GHS07
Storage Temperature:Omgivande
CAS Number:5326-23-8
Formula:C₆H₄ClNO₂
MDL Number:MFCD00006241