Biohazard Transport Carriers
Catalog # 216-4305
Supplier: KARTELL
Specifications
- Description:Safety box for tubes and containers
- W×D×H:330×175×180 mm
- Pk:1
Specifications
About this item
Transparent PC, silicone gasket and stainless steel handle.
- Completely autoclavable
- Biohazard symbol on the box
- User sheet with instructions approved by WHO
To carry tubes or urine stool containers under safe conditions.