Order Entry
Sweden
ContactUsLinkComponent
3,5-Dimethoxyaniline 98%
3,5-Dimethoxyaniline 98%
Certifikat
undefined
3,5-Dimethoxyaniline 98%

Specifikationstestresultat

  • Appearance:
    White to beige to grey to light brown Powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    54 to 57 °C
  • GC:
    ≥97.5 %

Specifikationstestresultat

Relaterad information