



Aniline for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:62-53-3
:
Formula:C₆H₅NH₂
MDL Number:MFCD00007629
:93.13 g/mol
Merck Index:13,00661
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6.2 °C
Density:1.019 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-539-3
UN:1547
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:76 °C
:GHS06
:GHS05
:GHS08
:GHS09
Specification Test Results
- Hill Formula:C₆H₇N
- Chemical formula:C₆H₅NH₂
- Molar Mass:93.13 g/mol
- Boiling point:184 °C (1013 hPa)
- Density:1.02 g/cm³ (20 °C)
- Explosion limit:1.2 - 11 %(V)
- Flash point:76 °C
- Ignition temperature:540 °C
- Melting Point:-6.2 °C
- pH value:8.8 (36 g/l, H₂O, 20 °C)
- Vapor pressure:0.5 hPa (20 °C)
- Refractive Index:1.5863 (20 °C, 589 nm)
- Solubility:36 g/l (20 °C)
- LD 50 dermal:LD50 Rabbit 840 mg/kg
- Storage class:6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
- WGK:WGK 2 water endangering
- R Phrase:R 23/24/25-40-41-43-48/23/24/25-68-50
- S Phrase:S 26-27-36/37/39-45-46-61-63
Specification Test Results
Related Information
Frequently Bought Together