Tubing, rubber
Supplier: KARTELL
Certificates
About this item
Natural rubber (NR), orange.
- Temperature resistance: −35 to +70 °C
Useful for gases, non-corrosive liquids and medium concentration acids.
Natural rubber (NR), orange.
Useful for gases, non-corrosive liquids and medium concentration acids.