Tubing, neoprene
Leverantör: KARTELL
Certifikat
Om denna produkt
Neoprene, black.
- No absorption of liquids
- Extremely flexible and durable
For normal laboratory use. Resistant to acids and solvents, benzole and similar excluded.
Temperature-resistance: –35 to +85 °C